4-[2-(4-Chloro-2-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049660-500MG 049660-2.5G
101888-540EA 364.62 USD
101888-540 101888-542
4-[2-(4-Chloro-2-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Chloro-2-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560062
CAS Number: 1219963-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR