3-[(4-Bromo-3-methylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049659-500MG 049659-2.5G
101888-536EA 364.62 USD
101888-536 101888-538
3-[(4-Bromo-3-methylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Bromo-3-methylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD13560061
CAS Number: 1219949-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR