3-[2-(4-Bromo-3-methylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049657-500MG 049657-2.5G
101888-528EA 364.62 USD
101888-528 101888-530
3-[2-(4-Bromo-3-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Bromo-3-methylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560059
CAS Number: 1219964-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR