4-[2-(4-Bromo-3-methylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049656-500MG
049656-2.5G
101888-524EA
364.62
USD
101888-524
101888-526
4-[2-(4-Bromo-3-methylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Bromo-3-methylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁BrClNO |
MDL Number:
MFCD13560058 CAS Number: 1219956-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...