2-Bromo-4-ethylphenyl-2-(2-piperidinyl)ethylether hydrochloride

Supplier: Matrix Scientific

049654-500MG 049654-2.5G
101888-516EA 364.62 USD
101888-516 101888-518
2-Bromo-4-ethylphenyl-2-(2-piperidinyl)ethylether hydrochloride
2-Bromo-4-ethylphenyl-2-(2-piperidinyl)ethylether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560056
CAS Number: 1219961-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR