2-Bromo-4-ethylphenyl-2-(3-piperidinyl)ethylether hydrochloride
Supplier: Matrix Scientific
049653-500MG
049653-2.5G
101888-512EA
364.62
USD
101888-512
101888-514
2-Bromo-4-ethylphenyl-2-(3-piperidinyl)ethylether hydrochloride
2-Bromo-4-ethylphenyl-2-(3-piperidinyl)ethylether hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₃BrClNO |
MDL Number:
MFCD13560055 CAS Number: 1219961-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...