2-Bromo-4-ethylphenyl-2-(4-piperidinyl)ethylether hydrochloride

Supplier: Matrix Scientific

049652-500MG 049652-2.5G
101888-508EA 364.62 USD
101888-508 101888-510
2-Bromo-4-ethylphenyl-2-(4-piperidinyl)ethylether hydrochloride
2-Bromo-4-ethylphenyl-2-(4-piperidinyl)ethylether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560054
CAS Number: 1219964-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR