2-Bromo-4-(tert-butyl)phenyl-3-pyrrolidinylmethylether hydrochloride

Supplier: Matrix Scientific

049651-500MG 049651-2.5G
101888-504EA 364.62 USD
101888-504 101888-506
2-Bromo-4-(tert-butyl)phenyl-3-pyrrolidinylmethylether hydrochloride
2-Bromo-4-(tert-butyl)phenyl-3-pyrrolidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MW: 348.72 g/mol
MDL Number: MFCD13560053
CAS Number: 1219948-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR