4-{2-[2-Bromo-4-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049648-500MG 049648-2.5G
101888-492EA 364.62 USD
101888-492 101888-494
4-{2-[2-Bromo-4-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[2-Bromo-4-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇BrClNO
MDL Number: MFCD13560050
CAS Number: 1220029-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR