2-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049646-500MG 049646-2.5G
101888-484EA 364.62 USD
101888-484 101888-486
2-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride
2-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃BrClNO
MDL Number: MFCD13560048
CAS Number: 1219961-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR