3-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
049645-500MG
049645-2.5G
101888-480EA
364.62
USD
101888-480
101888-482
3-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₉H₂₃BrClNO |
MDL Number:
MFCD13560047 CAS Number: 1220029-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...