4-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049644-500MG 049644-2.5G
101888-476EA 364.62 USD
101888-476 101888-478
4-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride
4-{2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃BrClNO
MDL Number: MFCD13560046
CAS Number: 1220016-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR