4-[2-(2-Bromo-4-isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049640-500MG 049640-2.5G
101888-460EA 364.62 USD
101888-460 101888-462
4-[2-(2-Bromo-4-isopropylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Bromo-4-isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560042
CAS Number: 1219948-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR