2-{2-[2,4-Di(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049638-500MG 049638-2.5G
101888-452EA 364.62 USD
101888-452 101888-454
2-{2-[2,4-Di(tert-butyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[2,4-Di(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₂₁H₃₆ClNO
MDL Number: MFCD13560040
CAS Number: 1219956-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR