4-{2-[2,4-Di(tert-butyl)phenoxy]ethyl}piperidine hydrochloride
Supplier: Matrix Scientific
049636-500MG
049636-2.5G
101888-444EA
364.62
USD
101888-444
101888-446
4-{2-[2,4-Di(tert-butyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[2,4-Di(tert-butyl)phenoxy]ethyl}piperidine hydrochloride
Hazard: Irritant
Formula:
C₂₁H₃₆ClNO |
MDL Number:
MFCD13560038 CAS Number: 1219961-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...