4-Bromo-2-(tert-butyl)phenyl-3-pyrrolidinylmethylether hydrochloride

Supplier: Matrix Scientific

049635-500MG 049635-2.5G
101888-440EA 364.62 USD
101888-440 101888-442
4-Bromo-2-(tert-butyl)phenyl-3-pyrrolidinylmethylether hydrochloride
4-Bromo-2-(tert-butyl)phenyl-3-pyrrolidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MW: 348.72 g/mol
MDL Number: MFCD13560037
CAS Number: 1219948-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR