3-{2-[4-Bromo-2-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049633-500MG 049633-2.5G
101888-432EA 364.62 USD
101888-432 101888-434
3-{2-[4-Bromo-2-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[4-Bromo-2-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇BrClNO
MDL Number: MFCD13560035
CAS Number: 1220027-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR