4-{2-[4-Bromo-2-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049632-500MG 049632-2.5G
101888-428EA 364.62 USD
101888-428 101888-430
4-{2-[4-Bromo-2-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[4-Bromo-2-(tert-butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇BrClNO
MDL Number: MFCD13560034
CAS Number: 1220029-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR