3-[(4-Bromo-2-isopropylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049631-500MG 049631-2.5G
101888-424EA 364.62 USD
101888-424 101888-426
3-[(4-Bromo-2-isopropylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Bromo-2-isopropylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560033
CAS Number: 1219980-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR