2-[2-(4-Bromo-2-isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049630-500MG 049630-2.5G
101888-420EA 364.62 USD
101888-420 101888-422
2-[2-(4-Bromo-2-isopropylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Bromo-2-isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560032
CAS Number: 1220027-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR