4-[2-(4-Bromo-2-isopropylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049628-500MG 049628-2.5G
101888-412EA 364.62 USD
101888-412 101888-414
4-[2-(4-Bromo-2-isopropylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Bromo-2-isopropylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560030
CAS Number: 1219982-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR