3-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049627-500MG 049627-2.5G
101888-408EA 364.62 USD
101888-408 101888-410
3-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-Bromo-4-(sec-butyl)phenoxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃BrClNO
MDL Number: MFCD13560029
CAS Number: 1219948-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR