3-{2-[2-Bromo-4-(sec-butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049625-500MG 049625-2.5G
101888-400EA 364.62 USD
101888-400 101888-402
3-{2-[2-Bromo-4-(sec-butyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[2-Bromo-4-(sec-butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇BrClNO
MDL Number: MFCD13560027
CAS Number: 1220029-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR