4-{2-[2-Bromo-4-(sec-butyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049624-500MG 049624-2.5G
101888-396EA 364.62 USD
101888-396 101888-398
4-{2-[2-Bromo-4-(sec-butyl)phenoxy]ethyl}piperidine hydrochloride
4-{2-[2-Bromo-4-(sec-butyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₇BrClNO
MDL Number: MFCD13560026
CAS Number: 1219956-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR