3-{[(1,6-Dibromo-2-naphthyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

049623-500MG 049623-2.5G
101888-392EA 364.62 USD
101888-392 101888-394
3-{[(1,6-Dibromo-2-naphthyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(1,6-Dibromo-2-naphthyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₆Br₂ClNO
MDL Number: MFCD13560025
CAS Number: 1220029-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR