2-{2-[(1,6-Dibromo-2-naphthyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049622-500MG 049622-2.5G
101888-388EA 364.62 USD
101888-388 101888-390
2-{2-[(1,6-Dibromo-2-naphthyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(1,6-Dibromo-2-naphthyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₀Br₂ClNO
MDL Number: MFCD13560024
CAS Number: 1219964-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR