2-{2-[(2,4-Dichloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

049618-500MG 049618-2.5G
101888-372EA 364.62 USD
101888-372 101888-374
2-{2-[(2,4-Dichloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2,4-Dichloro-1-naphthyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₀Cl₃NO
MW: 360.71 g/mol
MDL Number: MFCD13560020
CAS Number: 1220029-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR