3-[2-(4-Bromo-2-propylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049613-500MG 049613-2.5G
101888-352EA 364.62 USD
101888-352 101888-354
3-[2-(4-Bromo-2-propylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Bromo-2-propylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560015
CAS Number: 1219948-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR