4-[2-(4-Bromo-2-propylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049612-500MG 049612-2.5G
101888-348EA 364.62 USD
101888-348 101888-350
4-[2-(4-Bromo-2-propylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Bromo-2-propylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₅BrClNO
MDL Number: MFCD13560014
CAS Number: 1219964-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR