2-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049610-500MG 049610-2.5G
101888-340EA 364.62 USD
101888-340 101888-342
2-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂BrCl₂NO
MDL Number: MFCD13560012
CAS Number: 1220029-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR