3-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049609-500MG 049609-2.5G
101888-336EA 364.62 USD
101888-336 101888-338
3-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂BrCl₂NO
MDL Number: MFCD13560011
CAS Number: 1219956-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR