4-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049608-500MG 049608-2.5G
101888-332EA 364.62 USD
101888-332 101888-334
4-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Bromo-4-chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂BrCl₂NO
MDL Number: MFCD13560010
CAS Number: 1219964-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR