2-[2-(4-Benzyl-2-bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049606-500MG 049606-2.5G
101888-324EA 364.62 USD
101888-324 101888-326
2-[2-(4-Benzyl-2-bromophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Benzyl-2-bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₅BrClNO
MDL Number: MFCD13560008
CAS Number: 1220028-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR