4-[2-(4-Benzyl-2-bromophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049604-500MG 049604-2.5G
101888-316EA 364.62 USD
101888-316 101888-318
4-[2-(4-Benzyl-2-bromophenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Benzyl-2-bromophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₅BrClNO
MDL Number: MFCD13560006
CAS Number: 1219982-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR