4-Bromo-2-ethylphenyl-2-(2-piperidinyl)ethylether hydrochloride
Supplier: Matrix Scientific
049602-500MG
049602-2.5G
101888-308EA
364.62
USD
101888-308
101888-310
4-Bromo-2-ethylphenyl-2-(2-piperidinyl)ethylether hydrochloride
4-Bromo-2-ethylphenyl-2-(2-piperidinyl)ethylether hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₃BrClNO |
MDL Number:
MFCD13560004 CAS Number: 1219980-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...