4-Bromo-2-ethylphenyl-2-(3-piperidinyl)ethylether hydrochloride
Supplier: Matrix Scientific
049601-500MG
049601-2.5G
101888-304EA
364.62
USD
101888-304
101888-306
4-Bromo-2-ethylphenyl-2-(3-piperidinyl)ethylether hydrochloride
4-Bromo-2-ethylphenyl-2-(3-piperidinyl)ethylether hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₃BrClNO |
MDL Number:
MFCD13560003 CAS Number: 1219963-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...