4-Bromo-2-ethylphenyl-2-(4-piperidinyl)ethylether hydrochloride
Supplier: Matrix Scientific
049600-500MG
049600-2.5G
101888-300EA
364.62
USD
101888-300
101888-302
4-Bromo-2-ethylphenyl-2-(4-piperidinyl)ethylether hydrochloride
4-Bromo-2-ethylphenyl-2-(4-piperidinyl)ethylether hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₃BrClNO MW: 348.72 g/mol |
MDL Number:
MFCD13560002 CAS Number: 1220028-79-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...