2-Bromo-4-methoxyphenyl-3-pyrrolidinylmethylether hydrochloride

Supplier: Matrix Scientific

049599-500MG 049599-2.5G
101888-296EA 364.62 USD
101888-296 101888-298
2-Bromo-4-methoxyphenyl-3-pyrrolidinylmethylether hydrochloride
2-Bromo-4-methoxyphenyl-3-pyrrolidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO₂
MDL Number: MFCD13560001
CAS Number: 1219956-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR