3-[2-(2-Bromo-4-methoxyphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049597-500MG 049597-2.5G
101888-288EA 364.62 USD
101888-288 101888-290
3-[2-(2-Bromo-4-methoxyphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Bromo-4-methoxyphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO₂
MDL Number: MFCD13559999
CAS Number: 1219964-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR