4-[2-(2-Bromo-4-methoxyphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049596-500MG
049596-2.5G
101888-284EA
364.62
USD
101888-284
101888-286
4-[2-(2-Bromo-4-methoxyphenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Bromo-4-methoxyphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁BrClNO₂ |
MDL Number:
MFCD13559998 CAS Number: 1219964-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...