3-(Cyclobutylmethoxy)azetidine trifluoroacetate

Supplier: Matrix Scientific

049594-500MG 049594-2.5G
101888-276EA 423.08 USD
101888-276 101888-278
3-(Cyclobutylmethoxy)azetidine trifluoroacetate
3-(Cyclobutylmethoxy)azetidine trifluoroacetate

Hazard: Irritant

Formula: C₈H₁₆ClNO
MDL Number: MFCD23159155
CAS Number: 1427460-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR