3-[2-Nitro-4-(trifluoromethyl)phenoxy]azetidine

Supplier: Matrix Scientific

049592-500MG 049592-2.5G
101888-272EA 364.62 USD
101888-272 101888-274
3-[2-Nitro-4-(trifluoromethyl)phenoxy]azetidine
3-[2-Nitro-4-(trifluoromethyl)phenoxy]azetidine

Hazard: Irritant

Formula: C₁₀H₉F₃N₂O₃
MDL Number: MFCD13559995
CAS Number: 1220028-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR