3-(2-Chloro-4-nitrophenoxy)azetidine

Supplier: Matrix Scientific

049591-500MG 049591-2.5G
101888-268EA 364.62 USD
101888-268 101888-270
3-(2-Chloro-4-nitrophenoxy)azetidine
3-(2-Chloro-4-nitrophenoxy)azetidine

Hazard: Irritant

Formula: C₉H₉ClN₂O₃
MDL Number: MFCD13559994
CAS Number: 1220029-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR