2-(3-Azetidinyloxy)-5-(trifluoromethyl)pyridine trifluoroacetate

Supplier: Matrix Scientific

049590-500MG 049590-2.5G
101888-264EA 186.15 USD
101888-264 101888-266
2-(3-Azetidinyloxy)-5-(trifluoromethyl)pyridine trifluoroacetate
2-(3-Azetidinyloxy)-5-(trifluoromethyl)pyridine trifluoroacetate

Hazard: Irritant

Formula: C₁₁H₁₀F₆N₂O₃
MDL Number: MFCD21605758
CAS Number: 1361115-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR