2-(3-Azetidinyloxy)-6-bromopyridine hydrochloride

Supplier: Matrix Scientific

049589-500MG 049589-2.5G
101888-260EA 364.62 USD
101888-260 101888-262
2-(3-Azetidinyloxy)-6-bromopyridine hydrochloride
2-(3-Azetidinyloxy)-6-bromopyridine hydrochloride

Hazard: Irritant

Formula: C₈H₁₀BrClN₂O
MDL Number: MFCD22199247
CAS Number: 1707367-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR