2-(3-Azetidinyloxy)-6-chloropyrazine hydrochloride
Supplier: Matrix Scientific
049587-500MG
049587-2.5G
101888-252EA
364.62
USD
101888-252
101888-254
2-(3-Azetidinyloxy)-6-chloropyrazine hydrochloride
2-(3-Azetidinyloxy)-6-chloropyrazine hydrochloride
Hazard: Irritant
Formula:
C₇H₉Cl₂N₃O |
MDL Number:
MFCD19442200 CAS Number: 1332528-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...