3-[(4-Methoxybenzyl)oxy]azetidine

Supplier: Matrix Scientific

049585-500MG 049585-2.5G
101888-244EA 307.69 USD
101888-244 101888-246
3-[(4-Methoxybenzyl)oxy]azetidine
3-[(4-Methoxybenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD13559989
CAS Number: 1219980-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR