3-[(3,5-Dimethoxybenzyl)oxy]azetidine trifluoroacetate

Supplier: Matrix Scientific

049584-500MG 049584-2.5G
101888-240EA 384.62 USD
101888-240 101888-242
3-[(3,5-Dimethoxybenzyl)oxy]azetidine trifluoroacetate
3-[(3,5-Dimethoxybenzyl)oxy]azetidine trifluoroacetate

Hazard: Irritant

Formula: C₁₄H₁₈F₃NO₅
MDL Number: MFCD21605757
CAS Number: 1361115-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR