3-[(3,5-Difluorobenzyl)oxy]azetidine

Supplier: Matrix Scientific

049583-500MG 049583-2.5G
101888-236EA 364.62 USD
101888-236 101888-238
3-[(3,5-Difluorobenzyl)oxy]azetidine
3-[(3,5-Difluorobenzyl)oxy]azetidine

Hazard: Irritant

Formula: C₁₀H₁₁F₂NO
MDL Number: MFCD11874828
CAS Number: 1121595-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR