3-[(2,6-Difluorobenzyl)oxy]azetidinetrifluoroacetate salt

Supplier: Matrix Scientific

049581-500MG 049581-2.5G
101888-228EA 364.62 USD
101888-228 101888-230
3-[(2,6-Difluorobenzyl)oxy]azetidinetrifluoroacetate salt
3-[(2,6-Difluorobenzyl)oxy]azetidinetrifluoroacetate salt

Hazard: Irritant

Formula: C₁₂H₁₂F₅NO₃
MDL Number: MFCD26142756
CAS Number: 1707566-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR